Methyl 3-Hydroxyphenylacetate 97%

Supplier: Apollo Scientific

OR911917-25G
APOSOR911917-25GEA 157 EUR
APOSOR911917-25G
Methyl 3-Hydroxyphenylacetate 97%
Methyl 3-Hydroxyphenylacetate
Formula: C₉H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00017660
CAS Number: 42058-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR