2-Phenoxyacetohydrazide 96%

Supplier: Apollo Scientific

OR912008-5G
APOSOR912008-5GEA 98.6 EUR
APOSOR912008-5G
2-Phenoxyacetohydrazide 96%
2-Phenoxyacetohydrazide
Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00297030
CAS Number: 4664-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR