(R)-1-N-CBZ-2-Methyl-piperazine 95%

Supplier: Apollo Scientific

OR912199-1G OR912199-250MG OR912199-5G
APOSOR912199-1GEA 39.3 EUR
APOSOR912199-1G APOSOR912199-250MG APOSOR912199-5G
(R)-1-N-CBZ-2-Methyl-piperazine 95%
(R)-1-N-CBZ-2-Methyl-piperazine
Formula: C₁₃H₁₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04115051
CAS Number: 923565-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR