3,5-Dibromobenzyl alcohol 98%

Supplier: Apollo Scientific

OR912688-25G
APOSOR912688-25GEA 133 EUR
APOSOR912688-25G
3,5-Dibromobenzyl alcohol 98%
3,5-Dibromobenzyl alcohol
Formula: C₇H₆Br₂O
Storage Temperature: Ambient
MDL Number: MFCD01632143
CAS Number: 145691-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR