(R)-1-(Pyridin-3-yl)ethan-1-amine dihydrochloride 97%

Supplier: Apollo Scientific

Synonyms: (R)-1-(Pyridin-3-yl)ethanamine dihydrochloride

OR913004-1G OR913004-250MG
APOSOR913004-1GEA 549 EUR
APOSOR913004-1G APOSOR913004-250MG
(R)-1-(Pyridin-3-yl)ethan-1-amine dihydrochloride 97%
(R)-1-(Pyridin-3-yl)ethan-1-amine dihydrochloride
Formula: C₇H₁₀N₂·2ClH
Storage Temperature: Ambient
MDL Number: MFCD14705582
CAS Number: 40154-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR