[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride 98%

Supplier: Apollo Scientific
Warning

OR916291-5G OR916291-1G
APOSOR916291-5GEA 220 EUR
APOSOR916291-5G APOSOR916291-1G
[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride 98%
[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride
Formula: C₂₇H₂₆Cl₂P₂Pd
Storage Temperature: Ambient
MDL Number: MFCD03844773
CAS Number: 59831-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR