Ethyl 2-thiazol-4-ylacetate 95%

Supplier: Apollo Scientific

OR916582-1G
APOSOR916582-1GEA 376 EUR
APOSOR916582-1G
Ethyl 2-thiazol-4-ylacetate 95%
Ethyl 2-thiazol-4-ylacetate
Formula: C₇H₉NO₂S
Storage Temperature: Ambient
MDL Number: MFCD03011394
CAS Number: 120155-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR