Methyl butyrylacetate 95%

Supplier: Apollo Scientific

Synonyms: Methyl 3-oxohexanoate

OR917137-500G
APOSOR917137-500GEA 201 EUR
APOSOR917137-500G
Methyl butyrylacetate 95%
Methyl butyrylacetate
Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040995
CAS Number: 30414-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR