(2-Chlorothiazol-5-yl)methylamine hydrochloride 98%

Supplier: Apollo Scientific

OR918146-1G OR918146-25G OR918146-5G
APOSOR918146-1GEA 109 EUR
APOSOR918146-1G APOSOR918146-25G APOSOR918146-5G
(2-Chlorothiazol-5-yl)methylamine hydrochloride 98%
(2-Chlorothiazol-5-yl)methylamine hydrochloride
Formula: C₄H₆Cl₂N₂S
Storage Temperature: Ambient
MDL Number: MFCD09039253
CAS Number: 153471-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR