Chloride ionophore I 97%

Supplier: Apollo Scientific

OR918293-1G
APOSOR918293-1GEA 140 EUR
APOSOR918293-1G
Chloride ionophore I 97%
Chloride ionophore I
Formula: C₄₄H₂₈ClMnN₄
Storage Temperature: Ambient
MDL Number: MFCD00012153
CAS Number: 32195-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR