2,5-Dimethoxy-4-methylbenzaldehyde 96%

Supplier: Apollo Scientific

OR918388-10G OR918388-1G OR918388-5G
APOSOR918388-10GEA 466 EUR
APOSOR918388-10G APOSOR918388-1G APOSOR918388-5G
2,5-Dimethoxy-4-methylbenzaldehyde 96%
2,5-Dimethoxy-4-methylbenzaldehyde
Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD02253192
CAS Number: 4925-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR