5-Ethylisoxazole-3-carboxylic acid 96%

Supplier: Apollo Scientific

OR919230-1G OR919230-250MG
APOSOR919230-1GEA 256 EUR
APOSOR919230-1G APOSOR919230-250MG
5-Ethylisoxazole-3-carboxylic acid 96%
5-Ethylisoxazole-3-carboxylic acid
Formula: C₆H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD05864544
CAS Number: 52320-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR