4-Aminobenzophenone 98%

Supplier: Apollo Scientific

OR919301-5G OR919301-100G
APOSOR919301-5GEA 0 EUR
APOSOR919301-5G APOSOR919301-100G
4-Aminobenzophenone 98%
4-Aminobenzophenone
Formula: C₁₃H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD00007895
CAS Number: 1137-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR