3-(Aminomethyl)-4,6-dimethyl-1,2-dihydropyridin-2-one 97%

Supplier: Apollo Scientific

OR921633-25G OR921633-5G
APOSOR921633-25GEA 179 EUR
APOSOR921633-25G APOSOR921633-5G
3-(Aminomethyl)-4,6-dimethyl-1,2-dihydropyridin-2-one 97%
3-(Aminomethyl)-4,6-dimethyl-1,2-dihydropyridin-2-one
Formula: C₈H₁₂N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD06213260
CAS Number: 771579-27-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR