4-Methylazepan-4-ol hydrochloride 95%

Supplier: Apollo Scientific

OR921765-1G
APOSOR921765-1GEA 487 EUR
APOSOR921765-1G
4-Methylazepan-4-ol hydrochloride 95%
4-Methylazepan-4-ol hydrochloride
Formula: C₇H₁₅NO·ClH
MW: 165.661 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21362373
CAS Number: 91774-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR