1-[2-(Allyloxy)phenyl]ethanamine 95%

Supplier: Apollo Scientific

OR922195-1G OR922195-250MG
APOSOR922195-1GEA 632 EUR
APOSOR922195-1G APOSOR922195-250MG
1-[2-(Allyloxy)phenyl]ethanamine 95%
1-[2-(Allyloxy)phenyl]ethanamine
Formula: C₁₁H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD12802686
CAS Number: 1184578-88-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR