Isobutyryl bromide 95%

Supplier: Apollo Scientific

OR922323-25G OR922323-5G
APOSOR922323-25GEA 502 EUR
APOSOR922323-25G APOSOR922323-5G
Isobutyryl bromide 95%
Isobutyryl bromide
Formula: C₄H₇BrO
Storage Temperature: Ambient
MDL Number: MFCD00059468
CAS Number: 2736-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR