Decahydro-2-naphthol (mixture of isomers) 95%

Supplier: Apollo Scientific

OR922613-100G OR922613-25G
APOSOR922613-100GEA 270 EUR
APOSOR922613-100G APOSOR922613-25G
Decahydro-2-naphthol (mixture of isomers) 95%
Decahydro-2-naphthol (mixture of isomers)
Formula: C₁₀H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD00004132
CAS Number: 825-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR