Benzyl 3-methyl-4-oxopiperidine-1-carboxylate

Supplier: Apollo Scientific
Warning

OR92275-1G
APOSOR92275-1GEA 42.2 EUR
APOSOR92275-1G
Benzyl 3-methyl-4-oxopiperidine-1-carboxylate
Benzyl 3-methyl-4-oxopiperidine-1-carboxylate
Formula: C₁₄H₁₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD11848402
CAS Number: 1010115-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR