4-Isobutoxy-2-butanone 95%

Supplier: Apollo Scientific

OR924228-10G OR924228-1G OR924228-5G
APOSOR924228-10GEA 609 EUR
APOSOR924228-10G APOSOR924228-1G APOSOR924228-5G
4-Isobutoxy-2-butanone 95%
4-Isobutoxy-2-butanone
Formula: C₈H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD00059348
CAS Number: 31576-33-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR