(S)-3-Methoxypiperidine hydrochloride 95%

Supplier: Apollo Scientific

OR924852-1G OR924852-5G
APOSOR924852-1GEA 270 EUR
APOSOR924852-1G APOSOR924852-5G
(S)-3-Methoxypiperidine hydrochloride 95%
(S)-3-Methoxypiperidine hydrochloride
Formula: C₆H₁₃NO·ClH
MW: 151.634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17015900
CAS Number: 688809-96-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR