2,5-Dimethoxytoluene 95%

Supplier: Apollo Scientific

OR925248-25G
APOSOR925248-25GEA 95.7 EUR
APOSOR925248-25G
2,5-Dimethoxytoluene 95%
2,5-Dimethoxytoluene
Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 218
Melting Pt: 15
Storage Temperature: Ambient
MDL Number: MFCD00041901
CAS Number: 24599-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR