trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate

Supplier: Apollo Scientific

Synonyms: 2,2',2'',2'''-(trans-Cyclohexane-1,2-diylbis(azanetriyl))tetraacetic acid hydrate, DCTA monohydrate, CDTA monohydrate, trans-Cyclohex-1,2-ylenediaminetetra acetic acid monohydrate, (1,2-Cyclohexylenedinitrilo)tetraacetic acid monohydrate, 2,2',2'',2'''-[trans-Cyclohexane-1,2-diylbis(azanetriyl)]tetraacetic acid hydrate, CHELATON 4, Titriplex® IV, trans-1,2-Cyclohexanediaminetetraacetic acid monohydrate, trans-Cyclohexane-1,2-dinitrilotetraacetic acid monohydrate

OR926084-25G OR926084-100G
APOSOR926084-25GEA 23.2 EUR
APOSOR926084-25G APOSOR926084-100G
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
Formula: C₁₄H₂₂N₂O₈·1H₂O
Melting Pt: 213…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00149243
CAS Number: 125572-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR