5-Chloroisoindoline-1,3-dione 95%

Supplier: Apollo Scientific

OR926828-1G OR926828-5G
APOSOR926828-1GEA 0 EUR
APOSOR926828-1G APOSOR926828-5G
5-Chloroisoindoline-1,3-dione 95%
5-Chloroisoindoline-1,3-dione
Formula: C8H4ClNO2
Storage Temperature: Ambient
MDL Number: MFCD08703319
CAS Number: 7147-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR