2-Methyl-4,6-bis[(n-octylthio)methyl]phenol 98%

Supplier: Apollo Scientific

Synonyms: 2-Methyl-4,6-bis(n-octylsulphanylmethyl)phenol, 2-Methyl-4,6-bis((octylthio)methyl)phenol

OR929882-100G OR929882-25G OR929882-5G OR929882-1G
APOSOR929882-100GEA 183 EUR
APOSOR929882-100G APOSOR929882-25G APOSOR929882-5G APOSOR929882-1G
2-Methyl-4,6-bis[(n-octylthio)methyl]phenol 98%
2-Methyl-4,6-bis[(n-octylthio)methyl]phenol
Formula: C₂₅H₄₄OS₂
MW: 424.75 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00134699
CAS Number: 110553-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR