[2-Methyl-1-(1-methyl-1H-imidazol-2-yl)propyl]amine 95%

Supplier: Apollo Scientific

OR929979-1G OR929979-250MG
APOSOR929979-1GEA 464 EUR
APOSOR929979-1G APOSOR929979-250MG
[2-Methyl-1-(1-methyl-1H-imidazol-2-yl)propyl]amine 95%
[2-Methyl-1-(1-methyl-1H-imidazol-2-yl)propyl]amine
Formula: C₈H₁₅N₃
Storage Temperature: Ambient
MDL Number: MFCD09261514
CAS Number: 927986-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR