3-(Pyridin-3-yl)propan-1-amine 95%

Supplier: Apollo Scientific

Synonyms: 3-(3-Pyridyl)propylamine

OR930832-1G OR930832-250MG
APOSOR930832-1GEA 229 EUR
APOSOR930832-1G APOSOR930832-250MG
3-(Pyridin-3-yl)propan-1-amine 95%
3-(Pyridin-3-yl)propan-1-amine
Formula: C₈H₁₂N₂
MW: 136.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09886637
CAS Number: 41038-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR