6-Chloroquinazolin-2-amine 95%

Supplier: Apollo Scientific

OR931131-1G OR931131-250MG
APOSOR931131-1GEA 841 EUR
APOSOR931131-1G APOSOR931131-250MG
6-Chloroquinazolin-2-amine 95%
6-Chloroquinazolin-2-amine
Formula: C₈H₆ClN₃
Storage Temperature: Ambient
MDL Number: MFCD07368133
CAS Number: 20028-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR