3-Chloro-2-methoxy-4-pyridineboronic acid

Supplier: Apollo Scientific

Synonyms: 3-Chloro-2-methoxypyridine-4-boronic acid, (3-Chloro-2-methoxypyridin-4-yl)boronic acid

OR9330-1G
APOSOR9330-1GEA 372 EUR
APOSOR9330-1G
3-Chloro-2-methoxy-4-pyridineboronic acid
3-Chloro-2-methoxy-4-pyridineboronic acid
Formula: C₆H₇BClNO₃
Melting Pt: 152…155 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06798255
CAS Number: 957060-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR