5-Norbornene-2,3-dimethanol 95%

Supplier: Apollo Scientific

Synonyms: 2,3-Bis(hydroxymethyl)-5-norbornene, Bicyclo[2.2.1]hept-5-ene-2,3-dimethanol

OR933395-1G OR933395-5G
APOSOR933395-1GEA 75.4 EUR
APOSOR933395-1G APOSOR933395-5G
5-Norbornene-2,3-dimethanol 95%
5-Norbornene-2,3-dimethanol
Formula: C₉H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00474435
CAS Number: 85-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR