N,N-Dimethylthioacetamide 95%

Supplier: Apollo Scientific

OR937293-1G OR937293-5G
APOSOR937293-1GEA 37.7 EUR
APOSOR937293-1G APOSOR937293-5G
N,N-Dimethylthioacetamide 95%
N,N-Dimethylthioacetamide
Formula: C₄H₉NS
Storage Temperature: Ambient
MDL Number: MFCD00022178
CAS Number: 631-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR