3-phenylimidazolidine-2,4-dione 95%

Supplier: Apollo Scientific

OR937596-1G OR937596-250MG
APOSOR937596-1GEA 149 EUR
APOSOR937596-1G APOSOR937596-250MG
3-phenylimidazolidine-2,4-dione 95%
3-phenylimidazolidine-2,4-dione
Formula: C₉H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00209486
CAS Number: 2221-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR