1-Methyl-DL-proline 97%

Supplier: Apollo Scientific

OR938983-1G OR938983-250MG OR938983-500MG
APOSOR938983-1GEA 654 EUR
APOSOR938983-1G APOSOR938983-250MG APOSOR938983-500MG
1-Methyl-DL-proline 97%
1-Methyl-DL-proline
Formula: C₆H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD01605583
CAS Number: 68078-09-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR