2-((4-Methoxybenzyl)oxy)acetic acid 95%

Supplier: Apollo Scientific

OR939102-5G
APOSOR939102-5GEA 55.1 EUR
APOSOR939102-5G
2-((4-Methoxybenzyl)oxy)acetic acid 95%
2-((4-Methoxybenzyl)oxy)acetic acid
Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861881
CAS Number: 88920-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR