(+)-Limonene 1,2-epoxide 95%

Supplier: Apollo Scientific

OR939871-100G OR939871-25G OR939871-10G
APOSOR939871-100GEA 401 EUR
APOSOR939871-100G APOSOR939871-25G APOSOR939871-10G
(+)-Limonene 1,2-epoxide 95%
(+)-Limonene 1,2-epoxide
Storage Temperature: Refrigerator
CAS Number: 203719-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR