5-Hydroxy-3-methylindole 95%

Supplier: Apollo Scientific

Synonyms: 3-Methyl-1H-indol-5-ol

OR940620-1G OR940620-250MG
APOSOR940620-1GEA 139 EUR
APOSOR940620-1G APOSOR940620-250MG
5-Hydroxy-3-methylindole 95%
5-Hydroxy-3-methylindole
Formula: C₉H₉NO
Storage Temperature: Ambient
MDL Number: MFCD03095210
CAS Number: 1125-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR