N-Methyl-2-nitrobenzylamine 97%

Supplier: Apollo Scientific

Synonyms: N-Methyl-1-(2-nitrophenyl)methanamine

OR941225-1G OR941225-5G
APOSOR941225-1GEA 85.7 EUR
APOSOR941225-1G APOSOR941225-5G
N-Methyl-2-nitrobenzylamine 97%
N-Methyl-2-nitrobenzylamine
Formula: C₈H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04039891
CAS Number: 56222-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR