4-Phenyl-2-methyl-3-butyn-2-ol 95%

Supplier: Apollo Scientific

Synonyms: 2-Methyl-4-phenylbut-3-yn-2-ol

OR942808-5G
APOSOR942808-5GEA 143 EUR
APOSOR942808-5G
4-Phenyl-2-methyl-3-butyn-2-ol 95%
4-Phenyl-2-methyl-3-butyn-2-ol
Formula: C₁₁H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD00041572
CAS Number: 1719-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR