2-(Methylamino)nicotinaldehyde 95+%

Supplier: Apollo Scientific
Warning

OR943095-250MG
APOSOR943095-250MGEA 60.9 EUR
APOSOR943095-250MG
2-(Methylamino)nicotinaldehyde 95+%
2-(Methylamino)nicotinaldehyde
Formula: C₇H₈N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD05863669
CAS Number: 32399-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR