2-Chloro-6-methylamino-benzoic acid methyl ester 95%

Supplier: Apollo Scientific

OR943100-1G OR943100-250MG
APOSOR943100-1GEA 278 EUR
APOSOR943100-1G APOSOR943100-250MG
2-Chloro-6-methylamino-benzoic acid methyl ester 95%
2-Chloro-6-methylamino-benzoic acid methyl ester
Formula: C₉H₁₀ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD24387373
CAS Number: 1379595-97-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR