4-Amino-6-chloro-1H-indazole 95+%

Supplier: Apollo Scientific
Warning

OR945030-100MG OR945030-250MG OR945030-1G
APOSOR945030-100MGEA 36.4 EUR
APOSOR945030-100MG APOSOR945030-250MG APOSOR945030-1G
4-Amino-6-chloro-1H-indazole 95+%
4-Amino-6-chloro-1H-indazole
Formula: C₇H₆ClN₃
Storage Temperature: Ambient
MDL Number: MFCD07781370
CAS Number: 885519-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR