(3-P-Tolyl-isoxazol-5-yl)-methanol 95%

Supplier: Apollo Scientific

OR946560-1G OR946560-250MG
APOSOR946560-1GEA 841 EUR
APOSOR946560-1G APOSOR946560-250MG
(3-P-Tolyl-isoxazol-5-yl)-methanol 95%
(3-P-Tolyl-isoxazol-5-yl)-methanol
Formula: C₁₁H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD06738486
CAS Number: 206055-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR