5-Methoxynaphthalen-1-ol 95%

Supplier: Apollo Scientific

OR947028-1G OR947028-250MG
APOSOR947028-1GEA 754 EUR
APOSOR947028-1G APOSOR947028-250MG
5-Methoxynaphthalen-1-ol 95%
5-Methoxynaphthalen-1-ol
Formula: C₁₁H₁₀O₂
MW: 174.196 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00512042
CAS Number: 3588-80-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR