2-(tert-Butoxycarbonyl)-2-azabicyclo[3.1.0]hexane-1-carboxylic acid

Supplier: Apollo Scientific
Warning

OR948823-250MG
APOSOR948823-250MGEA 364 EUR
APOSOR948823-250MG
2-(tert-Butoxycarbonyl)-2-azabicyclo[3.1.0]hexane-1-carboxylic acid
2-(tert-Butoxycarbonyl)-2-azabicyclo[3.1.0]hexane-1-carboxylic acid
Formula: C₁₁H₁₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD17016465
CAS Number: 1251004-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR