2-Methylazepane hydrochloride 95%

Supplier: Apollo Scientific
Danger

OR950677-1G
APOSOR950677-1GEA 509 EUR
APOSOR950677-1G
2-Methylazepane hydrochloride 95%
2-Methylazepane hydrochloride
Formula: C₇H₁₅N·ClH
MW: 149.662 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27500876
CAS Number: 30796-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR