Phenyl propionate 95%

Supplier: Apollo Scientific

OR950919-25G OR950919-5G
APOSOR950919-25GEA 120 EUR
APOSOR950919-25G APOSOR950919-5G
Phenyl propionate 95%
Phenyl propionate
Formula: C₉H₁₀O₂
Boiling Pt: 211 °C
Density: 1,05 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00053765
CAS Number: 637-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR