4,7-Dimethoxy-1-indanone 95%

Supplier: Apollo Scientific

OR952308-5G
APOSOR952308-5GEA 133 EUR
APOSOR952308-5G
4,7-Dimethoxy-1-indanone 95%
4,7-Dimethoxy-1-indanone
Formula: C₁₁H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00757244
CAS Number: 52428-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR