6-Methoxyisoquinolin-1(2H)-one 95%

Supplier: Apollo Scientific

OR952438-1G
APOSOR952438-1GEA 143 EUR
APOSOR952438-1G
6-Methoxyisoquinolin-1(2H)-one 95%
6-Methoxyisoquinolin-1(2H)-one
Formula: C₁₀H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD12405870
CAS Number: 26829-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR