Methyl-1-aminocyclopropane-1-carboxylate 96%

Supplier: Apollo Scientific

Synonyms: Methyl 1-aminocyclopropanecarboxylate

OR952598-5G
APOSOR952598-5GEA 84.1 EUR
APOSOR952598-5G
Methyl-1-aminocyclopropane-1-carboxylate 96%
Methyl-1-aminocyclopropane-1-carboxylate
Formula: C₅H₉NO₂
Storage Temperature: Freezer
MDL Number: MFCD00906963
CAS Number: 72784-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR