2-(O-Tolylamino)ethanol 95%

Supplier: Apollo Scientific

OR952803-1G OR952803-250MG
APOSOR952803-1GEA 109 EUR
APOSOR952803-1G APOSOR952803-250MG
2-(O-Tolylamino)ethanol 95%
2-(O-Tolylamino)ethanol
Formula: C₉H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD00083105
CAS Number: 136-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR